missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1120036

 View more versions of this product

Catalog No. AAA1120036


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4'-Ethoxyacetanilide
134°C to 136°C
MFCD00009094
1869238
phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin
CCOC1=CC=C(C=C1)NC(=O)C
179.219
CHEBI:8050
97%
62-44-2
C10H13NO2
500 g
14,7204
CPJSUEIXXCENMM-UHFFFAOYSA-N
N-(4-ethoxyphenyl)acetamide
4754
179.22

Chemical Identifiers

62-44-2
179.219
CPJSUEIXXCENMM-UHFFFAOYSA-N
4754
N-(4-ethoxyphenyl)acetamide
C10H13NO2
MFCD00009094
phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin
CHEBI:8050
CCOC1=CC=C(C=C1)NC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H350

EINECSNumber : 200-533-0

RTECSNumber : AM4375000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only