missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Ethoxy-3'-(trifluoromethyl)acetophenone, 97%

Manufacturer:  Alfa Aesar™ H3444803

 View more versions of this product

Catalog No. AAH3444803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4'-Ethoxy-3'-(trifluoromethyl)acetophenone
851263-13-3
C11H11F3O2
KWNZFVILRICPQN-UHFFFAOYSA-N
1-[4-ethoxy-3-(trifluoromethyl)phenyl]ethanone
57361403
Solid
1g
97%
MFCD13196590
CCOC1=C(C=C(C=C1)C(=O)C)C(F)(F)F
232.202
232.2

Chemical Identifiers

851263-13-3
232.202
KWNZFVILRICPQN-UHFFFAOYSA-N
C11H11F3O2
MFCD13196590
57361403
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents