Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-3-nitropyridine hydrochloride, 97%

$56.36 - $56.36

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-3-nitropyridine hydrochloride
CAS 94602-04-7
Molecular Formula C7H8N2O3·HCl
Formula Weight 204.61
MDL Number MFCD01075663
Beilstein 3712456
Synonym 3-Nitro-4-ethoxypyridine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1474306

  SDS
alfa aesar™
L1474306
5g Each for $56.36
Description & Specifications

Specifications

Formula Weight 204.61
Chemical Name or Material 4-Ethoxy-3-nitropyridine hydrochloride
Synonym 3-Nitro-4-ethoxypyridine hydrochloride
Beilstein 3712456
MDL Number MFCD01075663
CAS 94602-04-7
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H8N2O3·HCl
Assay 97%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈270°C

Applications
4-Ethoxy-3-nitropyridine hydrochloride acts as an intermediate used in organic synthesis. It is also used in the preparation of 3,4-diaminopyridine.

Solubility
Soluble in water.

Notes
Hygroscopic and moisture sensitive.Incompatible with oxidizing agent and strong reducing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.