Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-3-methoxybenzyl alcohol

$159.04 - $672.61

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-3-methoxybenzyl alcohol
CAS 61813-58-9
Molecular Formula C10H14O3
Formula Weight 182.22g/mol
MDL Number MFCD00004656
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5034303

 
alfa aesar™
H50343-03
Each for $159.04

AAH5034306

 
alfa aesar™
H50343-06
Each for $672.61
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 4-Ethoxy-3-methoxybenzyl alcohol
CAS 61813-58-9
Molecular Formula C10H14O3
Formula Weight 182.22g/mol
MDL Number MFCD00004656