Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-3-methoxybenzaldehyde, 98+%

$69.46 - $239.58

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-3-methoxybenzaldehyde
CAS 120-25-2
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00016614
Beilstein 391169
Synonym 4-Ethoxy-m-anisaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1089714

  SDS
alfa aesar™
A1089714
25g Each for $69.46

AAA1089722

  SDS
alfa aesar™
A1089722
100g Each for $239.58
Description & Specifications

Specifications

Formula Weight 180.2
Chemical Name or Material 4-Ethoxy-3-methoxybenzaldehyde
TSCA TSCA
Assay ≥98%
Molecular Formula C10H12O3
MDL Number MFCD00016614
CAS 120-25-2
Beilstein 391169
Synonym 4-Ethoxy-m-anisaldehyde
Melting Point 59° to 63°C
Quantity 25g