Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-3,5-difluorobenzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-3,5-difluorobenzaldehyde
Molecular Formula C9H8F2O2
Formula Weight 186.16
MDL Number MFCD06247325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3267106

  SDS
alfa aesar™
H3267106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.16
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Chemical Name or Material 4-Ethoxy-3,5-difluorobenzaldehyde
Melting Point 27° to 29°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C9H8F2O2
Quantity 5g
MDL Number MFCD06247325
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.