Please login to your online account to display your discounted pricing

4-Ethoxy-2-nitroaniline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-2-nitroaniline
Name Note 97%
CAS 616-86-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H10N2O3
Linear Formula C2H5OC6H3(NO2)NH2
Formula Weight 182.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC184100010

 
thermo fisher scientific
AC184100010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 182.18
Physical Form Crystals or Crystalline Powder
Quantity 1g
Color Red to rust-brown
Melting Point 110° to 114°C
Beilstein 13, 521
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula C2H5OC6H3(NO2)NH2
Molecular Formula C8H10N2O3
MDL Number MFCD00007153
Assay Percent Range 97%
Chemical Name or Material 4-Ethoxy-2-nitroaniline
CAS 616-86-4
Synonym 2-Nitro-p-phenetidine
Name Note 97%