Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-2,6-difluorobenzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-2,6-difluorobenzaldehyde
Molecular Formula C9H8F2O2
Formula Weight 186.16
MDL Number MFCD09258695
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3175303

  SDS
alfa aesar™
H3175303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3175306

  SDS
alfa aesar™
H3175306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.16
MDL Number MFCD09258695
Melting Point 70° to 72°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Chemical Name or Material 4-Ethoxy-2,6-difluorobenzaldehyde
Molecular Formula C9H8F2O2
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.