Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-2,3-difluorophenol, 97%

$136.23 - $549.53

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-2,3-difluorophenol
CAS 126163-56-2
Molecular Formula C8H8F2O2
Formula Weight 174.14
MDL Number MFCD07368025
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3202903

  SDS
alfa aesar™
H3202903
1g Each for $136.23

AAH3202906

  SDS
alfa aesar™
H3202906
5g Each for $549.53
Description & Specifications

Specifications

Formula Weight 174.14
Chemical Name or Material 4-Ethoxy-2,3-difluorophenol
Assay 97%
Molecular Formula C8H8F2O2
MDL Number MFCD07368025
CAS 126163-56-2
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 65° to 77°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.