Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-2,3-difluorobenzyl alcohol, 97%

$87.43 - $347.88

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-2,3-difluorobenzyl alcohol
CAS 293299-94-2
Molecular Formula C9H10F2O2
Formula Weight 188.17
MDL Number MFCD09258704
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3286803

  SDS
alfa aesar™
H3286803
1g Each for $87.43

AAH3286806

  SDS
alfa aesar™
H3286806
5g Each for $347.88
Description & Specifications

Specifications

Formula Weight 188.17
Chemical Name or Material 4-Ethoxy-2,3-difluorobenzyl alcohol
Assay 97%
Molecular Formula C9H10F2O2
MDL Number MFCD09258704
CAS 293299-94-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 52° to 56°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.