Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-2,3-difluorobenzoic acid, 98%

$106.66 - $731.16

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-2,3-difluorobenzoic acid
CAS 124728-45-6
Molecular Formula C9H8F2O3
Formula Weight 202.15
MDL Number MFCD02258849
Synonym 2,3-Difluoro-4-ethoxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2608903

 
alfa aesar™
H2608903
1g Each for $264.76

AAH2608906

 
alfa aesar™
H2608906
5g Each for $731.16

AAH26089MD

 
alfa aesar™
H26089MD
250mg Each for $106.66
Description & Specifications

Specifications

Formula Weight 202.15
Chemical Name or Material 4-Ethoxy-2,3-difluorobenzoic acid
Assay 98%
Molecular Formula C9H8F2O3
MDL Number MFCD02258849
CAS 124728-45-6
Synonym 2,3-Difluoro-4-ethoxybenzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 200° to 203°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.