Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-2,3-difluorobenzaldehyde, 97%

$213.96 - $213.96

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-2,3-difluorobenzaldehyde
Molecular Formula C9H8F2O2
Formula Weight 186.16
MDL Number MFCD09258700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3245006

  SDS
alfa aesar™
H3245006
5g Each for $213.96
Description & Specifications

Specifications

Formula Weight 186.16
Chemical Name or Material 4-Ethoxy-2,3-difluorobenzaldehyde
Assay 97%
Molecular Formula C9H8F2O2
MDL Number MFCD09258700
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 70° to 73°C
Quantity 5g

Applications
4-Ethoxy-2,3-difluorobenzaldehyde, is used as a pharmaceutical intermediate in antibiotics, as sedatives, and for cancer treatment.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.