Learn More
4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 435360050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Ethoxy-1, 1, 1-trifluoro-3-buten-2-one | |
96.0 | |
1.1800g/mL | |
51°C | |
96% min. (GC) | |
MFCD00192131 | |
1.18 | |
YKYIFUROKBDHCY-UHFFFAOYSA-N | |
(E)-4-ethoxy-1,1,1-trifluorobut-3-en-2-one | |
(0.5% BHT stabilizer) | |
168.11 |
17129-06-5 | |
100.0 | |
51.0°C to 53.0°C (12.0mmHg) | |
Authentic | |
C6H7F3O2 | |
5g | |
4-ethoxy-1,1,1-trifluoro-3-buten-2-one, 4-ethoxy-1,1,1-trifluorobut-3-en-2-one, e-4-ethoxy-1,1,1-trifluorobut-3-en-2-one, 3e-4-ethoxy-1,1,1-trifluorobut-3-en-2-one, etfbo, 2-ethoxyvinyl trifluoromethyl ketone, e-4-ethoxy-1,1,1-trifluoro-3-buten-2-one, 3-buten-2-one, 4-ethoxy-1,1,1-trifluoro, 4-ethoxy, 4-ethoxy-1,1,1-trifluoro-but-3-en-2-one | |
CCOC=CC(=O)C(F)(F)F | |
168.12 | |
5709222 | |
97% |
Chemical Identifiers
17129-06-5 | |
168.12 | |
YKYIFUROKBDHCY-UHFFFAOYSA-N | |
5709222 | |
CCOC=CC(=O)C(F)(F)F |
C6H7F3O2 | |
MFCD00192131 | |
4-ethoxy-1,1,1-trifluoro-3-buten-2-one, 4-ethoxy-1,1,1-trifluorobut-3-en-2-one, e-4-ethoxy-1,1,1-trifluorobut-3-en-2-one, 3e-4-ethoxy-1,1,1-trifluorobut-3-en-2-one, etfbo, 2-ethoxyvinyl trifluoromethyl ketone, e-4-ethoxy-1,1,1-trifluoro-3-buten-2-one, 3-buten-2-one, 4-ethoxy-1,1,1-trifluoro, 4-ethoxy, 4-ethoxy-1,1,1-trifluoro-but-3-en-2-one | |
(E)-4-ethoxy-1,1,1-trifluorobut-3-en-2-one |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
RUO â Research Use Only