Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, 94%, stab.

Chemical Identifiers

Chemical Name or Material 4-Ethoxy-1,1,1-trifluoro-3-buten-2-one
CAS 17129-06-5
Name Note stabilized
Molecular Formula C6H7F3O2
Formula Weight 168.13
MDL Number MFCD00192131
UN Number UN3384
Synonym ETFBO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2697503

  SDS
alfa aesar™
H2697503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2697506

  SDS
alfa aesar™
H2697506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.13
Odor Ether-like
Health Hazard 2 GHS H Statement
H330-H372-H226-H302-H315-H319-H317
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
UN Number UN3384
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
Molecular Formula C6H7F3O2
Boiling Point 159°C
Synonym ETFBO
Flash Point 52°C (126°F)
Chemical Name or Material 4-Ethoxy-1,1,1-trifluoro-3-buten-2-one
Name Note stabilized
Quantity 1g
Health Hazard 3 GHS P Statement
P280h-P273-P305+P351+P338-P304+P340-P309-P310
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Melting Point −30°C
Assay 94%
Recommended Storage −30° to −10°C
CAS 17129-06-5
Health Hazard 1 Danger
MDL Number MFCD00192131
Safety and Handling

Danger

GHS P Statement
P280h-P273-P305+P351+P338-P304+P340-P309-P310
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H330-H372-H226-H302-H315-H319-H317
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

−30° to −10°C