missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Dodecyl-o-cresol 98.0+%, TCI America™
Supplier: TCI America D230410G
Specifications
4-Dodecyl-o-cresol | |
White-Yellow | |
MFCD00191639 | |
4-Lauryl-o-cresol | |
CCCCCCCCCCCCC1=CC(C)=C(O)C=C1 | |
276.46 | |
276.46 | |
Crystals |
29665-59-6 | |
C19H32O | |
10 g | |
RJLWSJUUDKAMPQ-UHFFFAOYSA-N | |
4-dodecyl-2-methylphenol | |
578742 | |
≥98.0% (GC) |
Chemical Identifiers
29665-59-6 | |
276.46 | |
RJLWSJUUDKAMPQ-UHFFFAOYSA-N | |
578742 | |
CCCCCCCCCCCCC1=CC(C)=C(O)C=C1 |
C19H32O | |
MFCD00191639 | |
4-Lauryl-o-cresol | |
4-dodecyl-2-methylphenol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator