Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dodecanone, 99%

$237.05 - $701.91

Chemical Identifiers

Chemical Name or Material 4-Dodecanone
CAS 6137-26-4
Molecular Formula C12H24O
Formula Weight 184.32
MDL Number MFCD00015300
Beilstein 1703138
Synonym n-Octyl n-propyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0419218

 
alfa aesar™
L0419218
50g Each for $237.05

AAL0419230

 
alfa aesar™
L0419230
250g Each for $701.91
Description & Specifications

Specifications

Formula Weight 184.32
MDL Number MFCD00015300
CAS 6137-26-4
Assay 99%
Molecular Formula C12H24O
Beilstein 1703138
Synonym n-Octyl n-propyl ketone
Boiling Point 65°C (0.25mmHg)
Quantity 50g
Chemical Name or Material 4-Dodecanone