Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dodecanone, 99%


Quantity 250g

Manufacturer: alfa aesar™  L0419230

 View more versions of this product

Catalog No. AAL0419230

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 4-Dodecanone
Quantity 250g
CAS 6137-26-4
Assay 99%
Molecular Formula C12H24O
Formula Weight 184.32
MDL Number MFCD00015300
Beilstein 1703138
Synonym n-Octyl n-propyl ketone
Boiling Point 65°C (0.25mmHg)

Specifications

Chemical Name or Material 4-Dodecanone
CAS 6137-26-4
Molecular Formula C12H24O
Formula Weight 184.32
MDL Number MFCD00015300
Beilstein 1703138
Synonym n-Octyl n-propyl ketone