missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dodecanone, 99%

Manufacturer:  Alfa Aesar™ L0419209

 View more versions of this product

Catalog No. AAL0419209



Specifications

Specifications

4-Dodecanone
10g
C12H24O
1703138
AVQSOIZWTINZLU-UHFFFAOYSA-N
dodecan-4-one
80226
99%
65°C (0.25mmHg)
6137-26-4
MFCD00015300
4-dodecanone, octyl propyl ketone, acmc-1bh4v
CCCCCCCCC(=O)CCC
184.323
184.32

Chemical Identifiers

6137-26-4
184.323
AVQSOIZWTINZLU-UHFFFAOYSA-N
80226
CCCCCCCCC(=O)CCC
C12H24O
MFCD00015300
4-dodecanone, octyl propyl ketone, acmc-1bh4v
dodecan-4-one
Documents