missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dodecanol, 99%

Manufacturer:  Alfa Aesar™ L0292106

 View more versions of this product

Catalog No. AAL0292106



Specifications

Specifications

4-Dodecanol
11°C to 12°C
10203-32-4
MFCD00014413
4-dodecanol
CCCCCCCCC(CCC)O
186.339
186.34
130°C to 132°C (19mmHg)
5g
C12H26O
1738597
ZGSIAHIBHSEKPB-UHFFFAOYSA-N
dodecan-4-ol
66291
99%

Chemical Identifiers

10203-32-4
186.339
ZGSIAHIBHSEKPB-UHFFFAOYSA-N
66291
CCCCCCCCC(CCC)O
C12H26O
MFCD00014413
4-dodecanol
dodecan-4-ol
Documents