Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dodecanol, 99%

$100.51 - $100.51

Chemical Identifiers

Chemical Name or Material 4-Dodecanol
CAS 10203-32-4
Molecular Formula C12H26O
Formula Weight 186.34
MDL Number MFCD00014413
Beilstein 1738597
Synonym n-Octyl-n-propyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0292106

 
alfa aesar™
L0292106
5g Each for $100.51
Description & Specifications

Specifications

Formula Weight 186.34
Chemical Name or Material 4-Dodecanol
CAS 10203-32-4
Assay 99%
Molecular Formula C12H26O
MDL Number MFCD00014413
Beilstein 1738597
Synonym n-Octyl-n-propyl carbinol
Boiling Point 130° to 132°C (19mmHg)
Melting Point 11° to 12°C
Quantity 5g