Please login to your online account to display your discounted pricing

4-[(Dipropylamino)sulfonyl]benzoic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-[(Dipropylamino)sulfonyl]benzoic acid
CAS 57-66-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H19NO4S
Linear Formula [CH3(CH2)2]2NSO2C6H4CO2H
Formula Weight 285.36
MDL Number MFCD00038402
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

SB00915EA

 
fisher scientific company
SB00915EA
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 285.36
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C13H19NO4S
Linear Formula [CH3(CH2)2]2NSO2C6H4CO2H
Chemical Name or Material 4-[(Dipropylamino)sulfonyl]benzoic acid
Assay Percent Range 97%
MDL Number MFCD00038402
Health Hazard 1 Warning
CAS 57-66-9
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.