Please login to your online account to display your discounted pricing

4-Diphenylaminobenzaldehyde 98%, ACROS Organics™

$165.86 - $165.86

Chemical Identifiers

Chemical Name or Material 4-Diphenylaminobenzaldehyde
CAS 4181-05-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C19H15NO
Linear Formula (C6H5)2NC6H4C(=O)H
Formula Weight 273.33
MDL Number MFCD00145131
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391180050

  SDS
thermo fisher scientific
AC391180050
Glass bottle 5g Each for $165.86

AC391180250

  SDS
thermo fisher scientific
AC391180250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 273.33
Quantity 5g
Color Pale yellow
Melting Point 129° to 133°C
CAS Max % 100.0
Chemical Name or Material 4-Diphenylaminobenzaldehyde
Infrared Spectrum Authentic
Molecular Formula C19H15NO
Linear Formula (C6H5)2NC6H4C(=O)H
MDL Number MFCD00145131
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 98%
CAS 4181-05-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.