missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dimethylcarbamoyl-3-fluorobenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53117MD

 View more versions of this product

Catalog No. AAH53117MD



Specifications

Specifications

4-Dimethylcarbamoyl-3-fluorobenzeneboronic acid
250mg
C9H11BFNO3
4-dimethylcarbamoyl-3-fluorophenylboronic acid, 4-dimethylcarbamoyl-3-fluorophenyl boronic acid, 4-dimethylcarbamoyl-3-fluorophenyl boronic acid, 4-dimethylcarbamoyl-3-fluorobenzeneboronic acid, acmc-209qm6, 4-dimethylcarbamoyl-3-fluorophenyl boronicacid, 4-dimethyl carbamoyl-3-fluorophenylboronic acid, 4-dimethylcarbamoyl-3-fluoranyl-phenyl boronic acid, 4-dimethylamino oxo methyl-3-fluorophenyl boronic acid, boronic acid,b-4-dimethylamino carbonyl-3-fluorophenyl
B(C1=CC(=C(C=C1)C(=O)N(C)C)F)(O)O
210.999
211
147°C to 149°C
874289-13-1
MFCD08436060
KEZPQOGKDGNRHH-UHFFFAOYSA-N
[4-(dimethylcarbamoyl)-3-fluorophenyl]boronic acid
44717474
98%

Chemical Identifiers

874289-13-1
210.999
KEZPQOGKDGNRHH-UHFFFAOYSA-N
44717474
B(C1=CC(=C(C=C1)C(=O)N(C)C)F)(O)O
C9H11BFNO3
MFCD08436060
4-dimethylcarbamoyl-3-fluorophenylboronic acid, 4-dimethylcarbamoyl-3-fluorophenyl boronic acid, 4-dimethylcarbamoyl-3-fluorophenyl boronic acid, 4-dimethylcarbamoyl-3-fluorobenzeneboronic acid, acmc-209qm6, 4-dimethylcarbamoyl-3-fluorophenyl boronicacid, 4-dimethyl carbamoyl-3-fluorophenylboronic acid, 4-dimethylcarbamoyl-3-fluoranyl-phenyl boronic acid, 4-dimethylamino oxo methyl-3-fluorophenyl boronic acid, boronic acid,b-4-dimethylamino carbonyl-3-fluorophenyl
[4-(dimethylcarbamoyl)-3-fluorophenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents