missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dimethylcarbamoyl-2-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H55993MD

 View more versions of this product

Catalog No. AAH55993MD



Specifications

Specifications

4-Dimethylcarbamoyl-2-fluorobenzeneboronic acid
874289-30-2
MFCD11855833
IOTCMPCBVDPZTB-UHFFFAOYSA-N
[4-(dimethylcarbamoyl)-2-fluorophenyl]boronic acid
46738645
97%
250mg
C9H11BFNO3
4-dimethyl carbamoyl-2-fluorophenylboronic acid
B(C1=C(C=C(C=C1)C(=O)N(C)C)F)(O)O
210.999
211

Chemical Identifiers

874289-30-2
210.999
IOTCMPCBVDPZTB-UHFFFAOYSA-N
46738645
B(C1=C(C=C(C=C1)C(=O)N(C)C)F)(O)O
C9H11BFNO3
MFCD11855833
4-dimethyl carbamoyl-2-fluorophenylboronic acid
[4-(dimethylcarbamoyl)-2-fluorophenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents