Learn More
4-Dimethylaminobutyric acid hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1661014
Description
4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.
Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water and oxidizing agents.
Specifications
4-Dimethylaminobutyric acid hydrochloride | |
153°C to 155°C | |
MFCD00012613 | |
Hygroscopic | |
RDTALXUBMCLWBB-UHFFFAOYSA-N | |
4-(dimethylamino)butanoic acid;hydrochloride | |
12691640 | |
98% |
69954-66-1 | |
C6H14ClNO2 | |
25 g | |
4-dimethylamino butanoic acid hydrochloride, 4-dimethylaminobutyric acid hydrochloride, 4-dimethylamino butyric acid hydrochloride, butanoic acid, 4-dimethylamino-, hydrochloride, 4-dimethylamino butanoic acid hcl, 4-dimethylamino butyric acid hcl salt, 4-dimethylamino-butyric acid hydrochloride, 4-dimethylamino-butyric acid hydrochloride salt | |
CN(C)CCCC(=O)O.Cl | |
167.633 | |
167.64 |
Chemical Identifiers
69954-66-1 | |
167.633 | |
RDTALXUBMCLWBB-UHFFFAOYSA-N | |
12691640 | |
CN(C)CCCC(=O)O.Cl |
C6H14ClNO2 | |
MFCD00012613 | |
4-dimethylamino butanoic acid hydrochloride, 4-dimethylaminobutyric acid hydrochloride, 4-dimethylamino butyric acid hydrochloride, butanoic acid, 4-dimethylamino-, hydrochloride, 4-dimethylamino butanoic acid hcl, 4-dimethylamino butyric acid hcl salt, 4-dimethylamino-butyric acid hydrochloride, 4-dimethylamino-butyric acid hydrochloride salt | |
4-(dimethylamino)butanoic acid;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only