Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dimethylaminobutyraldehyde diethyl acetal, 97%

$55.89 - $563.53

Chemical Identifiers

Chemical Name or Material 4-Dimethylaminobutyraldehyde diethyl acetal
CAS 1116-77-4
Molecular Formula (CH3)2N(CH2)3CH(OCH2CH3)2
Formula Weight 189.3
MDL Number MFCD00671479
Synonym 4,4-Diethoxy-N,N-dimethyl-1-butylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2468614

  SDS
alfa aesar™
B2468614
25g Each for $55.89

AAB2468636

  SDS
alfa aesar™
B2468636
500g Each for $563.53
Description & Specifications

Specifications

Formula Weight 189.3
Chemical Name or Material 4-Dimethylaminobutyraldehyde diethyl acetal
Assay 97%
Molecular Formula (CH3)2N(CH2)3CH(OCH2CH3)2
MDL Number MFCD00671479
CAS 1116-77-4
Quantity 25g
Flash Point 70°C (158°F)
Synonym 4,4-Diethoxy-N,N-dimethyl-1-butylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.