Learn More
4-Dimethylaminobenzoic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 115760250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Dimethylaminobenzoic acid | |
97.5 | |
170g/mL | |
Authentic | |
Plastic bottle | |
(CH3)2NC6H4CO2H | |
25g | |
15, 3254 | |
Solubility in water: .. Other solubilities: soluble in hcl and koh solutions,soluble in alcohol,sparingly soluble in ether,practically insoluble in acetic acid | |
CN(C)C1=CC=C(C=C1)C(=O)O | |
165.19 | |
165.19 |
619-84-1 | |
100.0 | |
170°C | |
98% | |
C9H11NO2 | |
MFCD00002537 | |
14, 426 | |
4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid | |
YDIYEOMDOWUDTJ-UHFFFAOYSA-N | |
4-(dimethylamino)benzoic acid | |
12092 | |
98% |
Chemical Identifiers
619-84-1 | |
165.19 | |
YDIYEOMDOWUDTJ-UHFFFAOYSA-N | |
12092 | |
CN(C)C1=CC=C(C=C1)C(=O)O |
C9H11NO2 | |
MFCD00002537 | |
4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid | |
4-(dimethylamino)benzoic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 210-615-8
RUO â Research Use Only