Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dimethylaminobenzoic acid, 98+%

$135.61 - $544.91

Chemical Identifiers

Chemical Name or Material 4-Dimethylaminobenzoic acid
CAS 619-84-1
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD00002537
Beilstein 386867
Merck Index 14,3232
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1433822

 
alfa aesar™
A1433822
100g Each for $135.61

AAA1433830

 
alfa aesar™
A1433830
250g Each for $294.00

AAA1433836

 
alfa aesar™
A1433836
500g Each for $544.91
Description & Specifications

Specifications

Formula Weight 165.19
Chemical Name or Material 4-Dimethylaminobenzoic acid
TSCA TSCA
Assay ≥98%
Molecular Formula C9H11NO2
Melting Point ≈240°C (dec.)
MDL Number MFCD00002537
CAS 619-84-1
Beilstein 386867
Quantity 100g
Flash Point 170°C (338°F)
Merck Index 14,3232
Odor Odorless
RTECS Number DG8735000

Applications
4-Dimethylaminobenzoic acid a substituted benzoic acid that acts as an inhibitor of ultraviolet mediated damage to skin. It is a biotransformation product of 2-ethylhexyl 4- (N,N-dimethylaminobenzo)benzoate(EDP), a widely used UV filters in sunscreen cosmetics and other cosmetic products.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.