missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Dimethylaminobenzaldehyde, Reagent ACS, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 423635000

 View more versions of this product

Catalog No. AC423635000


 

Specifications Description & Specifications

Specifications

Crystalline Powder
149.19
Light yellow to slightly green
01,273; 02,146
73° to 75°C
Solubility in water: 0.3g/L (20°C) Other solubilities: Soluble in acetic acid, Soluble in alcohol, ether and chloroform
MFCD00003381
Authentic
100-10-7
4-Dimethylaminobenzaldehyde
ACS Reagent Grade
500g
14, 31
15, 3252
5.15
Glass bottle
Ehrlich's Reagent
0.1% max.
(CH3)2NC6H4CHO
C9H11NO

Chemical Identifiers

100-10-7
(CH3)2NC6H4CHO
MFCD00003381
4-Dimethylaminobenzaldehyde
C9H11NO
149.19
Ehrlich's Reagent
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
Harmful to aquatic life with long lasting effects. Harmful if swallowed.

GHS P Statement
Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Warning

EINECSNumber : 202-819-0

RTECSNumber : CU5775000

TSCA : TSCA