missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Dimethylaminobenzaldehyde, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 168760250

 View more versions of this product

Catalog No. AC168760250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Dimethylaminobenzaldehyde
70°C to 75°C
176°C to 177°C (17.0 mmHg)
Authentic
Plastic bottle
(CH3)2NC6H4CHO
25 g
01,273; 02,146
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-(dimethylamino)benzaldehyde
7479
99+%
100-10-7
Green to Yellow
164°C
99% min. (GC)
C9H11NO
MFCD00003381
14,31
15,3252
Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in acetic acid,soluble in alcohol,ether and chloroform
CN(C)C1=CC=C(C=O)C=C1
149.19
149.19
Crystalline Powder

Chemical Identifiers

100-10-7
149.19
BGNGWHSBYQYVRX-UHFFFAOYSA-N
7479
CN(C)C1=CC=C(C=O)C=C1
C9H11NO
MFCD00003381
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
4-(dimethylamino)benzaldehyde
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 202-819-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only