missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dimethylaminobenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ A1171236

 View more versions of this product

Catalog No. AAA1171236



Description

Description

Applications
4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

Solubility
Soluble in alcohol, ether, chloroform, acetic acid. Insoluble in water.

Notes
Light sensitive. Stable at room temperature.
Specifications

Specifications

4-Dimethylaminobenzaldehyde
Strong
73°C to 75°C
100-10-7
MFCD00003381
14,3230
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-(dimethylamino)benzaldehyde
7479
98%
164°C (327°F)
176°C to 177°C (17mmHg)
500g
C9H11NO
606802
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
CN(C)C1=CC=C(C=C1)C=O
149.193
149.19

Chemical Identifiers

100-10-7
149.193
BGNGWHSBYQYVRX-UHFFFAOYSA-N
7479
CN(C)C1=CC=C(C=C1)C=O
C9H11NO
MFCD00003381
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
4-(dimethylamino)benzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CU5775000

TSCA : TSCA

SDS Documents