Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dimethylaminoazobenzene-4'-isothiocyanate, 97%

$78.81 - $247.82

Chemical Identifiers

Chemical Name or Material 4-Dimethylaminoazobenzene-4'-isothiocyanate
CAS 7612-98-8
Molecular Formula C15H14N4S
Formula Weight 282.37
MDL Number MFCD00004812
Beilstein 752568
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1258103

  SDS
alfa aesar™
A1258103
1g Each for $247.82

AAA1258101

  SDS
alfa aesar™
A1258101
0.25g Each for $78.81
Description & Specifications

Specifications

Formula Weight 282.37
Chemical Name or Material 4-Dimethylaminoazobenzene-4'-isothiocyanate
Assay 97%
Molecular Formula C15H14N4S
Melting Point 169° to 171°C
MDL Number MFCD00004812
CAS 7612-98-8
Beilstein 752568
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NX9125000
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.