missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Dimethylamino)pyridine tribromide, 97%

Manufacturer:  Alfa Aesar™ H5501022

 View more versions of this product

Catalog No. AAH5501022



Specifications

Specifications

4-(Dimethylamino)pyridine tribromide
100g
C7H10Br3N2-
UN3261
TYDZHQMKSLAFKM-UHFFFAOYSA-N
361.883
362.89
129°C to 132°C
92976-81-3
MFCD00043439
4-dimethylamino pyridine hydrobromide perbromide
CN(C)C1=CC=NC=C1.Br[Br-]Br
131863682
97%

Chemical Identifiers

92976-81-3
361.883
TYDZHQMKSLAFKM-UHFFFAOYSA-N
131863682
C7H10Br3N2-
MFCD00043439
4-dimethylamino pyridine hydrobromide perbromide
CN(C)C1=CC=NC=C1.Br[Br-]Br
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents