missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Dimethylamino)pyridine tribromide, 97%

Manufacturer:  Alfa Aesar™ H5501006

 View more versions of this product

Catalog No. AAH5501006



Specifications

Specifications

4-(Dimethylamino)pyridine tribromide
5g
C7H10Br3N2-
UN3261
TYDZHQMKSLAFKM-UHFFFAOYSA-N
361.883
362.89
129°C to 132°C
92976-81-3
MFCD00043439
4-dimethylamino pyridine hydrobromide perbromide
CN(C)C1=CC=NC=C1.Br[Br-]Br
131863682
97%

Chemical Identifiers

92976-81-3
361.883
TYDZHQMKSLAFKM-UHFFFAOYSA-N
131863682
C7H10Br3N2-
MFCD00043439
4-dimethylamino pyridine hydrobromide perbromide
CN(C)C1=CC=NC=C1.Br[Br-]Br
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents