Learn More
4-(Dimethylamino)piperidine, 97%, Thermo Scientific Chemicals
$262.58 - $951.03
Chemical Identifiers
CAS | 50533-97-6 |
---|---|
Molecular Formula | C7H16N2 |
Molecular Weight (g/mol) | 128.22 |
MDL Number | MFCD00023144 |
InChI Key | YFJAIURZMRJPDB-UHFFFAOYSA-N |
Synonym | 4-dimethylamino piperidine, 4-dimethylaminopiperidine, 4-n,n-dimethylamino piperidine, 4-piperidinamine, n,n-dimethyl, 4-dimethylamino-piperidine, dimethyl-piperidin-4-yl-amine, pubchem6749, acmc-1akc4, dimethylpiperidin-4-ylamine |
PubChem CID | 417391 |
IUPAC Name | N,N-dimethylpiperidin-4-amine |
SMILES | CN(C)C1CCNCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2017606
|
Thermo Scientific Chemicals
L2017606 |
5 g |
Each for $262.58
|
|
|||||
AAL2017614
|
Thermo Scientific Chemicals
L2017614 |
25 g |
Each for $951.03
|
|
|||||
Description
4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.
Notes
Incompatible with air, water, oxidizing agents and heat.
Chemical Identifiers
50533-97-6 | |
128.22 | |
YFJAIURZMRJPDB-UHFFFAOYSA-N | |
417391 | |
CN(C)C1CCNCC1 |
C7H16N2 | |
MFCD00023144 | |
4-dimethylamino piperidine, 4-dimethylaminopiperidine, 4-n,n-dimethylamino piperidine, 4-piperidinamine, n,n-dimethyl, 4-dimethylamino-piperidine, dimethyl-piperidin-4-yl-amine, pubchem6749, acmc-1akc4, dimethylpiperidin-4-ylamine | |
N,N-dimethylpiperidin-4-amine |
Specifications
50533-97-6 | |
C7H16N2 | |
MFCD00023144 | |
UN2735 | |
4-dimethylamino piperidine, 4-dimethylaminopiperidine, 4-n,n-dimethylamino piperidine, 4-piperidinamine, n,n-dimethyl, 4-dimethylamino-piperidine, dimethyl-piperidin-4-yl-amine, pubchem6749, acmc-1akc4, dimethylpiperidin-4-ylamine | |
CN(C)C1CCNCC1 | |
128.22 | |
128.22 | |
4-(Dimethylamino)piperidine |
187°C | |
1.476 | |
5 g | |
1190 | |
YFJAIURZMRJPDB-UHFFFAOYSA-N | |
N,N-dimethylpiperidin-4-amine | |
417391 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 256-617-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only