Learn More
4-(Dimethylamino)piperidine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L2017606
Description
4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.
Notes
Incompatible with air, water, oxidizing agents and heat.
Specifications
4-(Dimethylamino)piperidine | |
187°C | |
1.476 | |
5 g | |
1190 | |
YFJAIURZMRJPDB-UHFFFAOYSA-N | |
N,N-dimethylpiperidin-4-amine | |
417391 | |
97% |
50533-97-6 | |
C7H16N2 | |
MFCD00023144 | |
UN2735 | |
4-dimethylamino piperidine, 4-dimethylaminopiperidine, 4-n,n-dimethylamino piperidine, 4-piperidinamine, n,n-dimethyl, 4-dimethylamino-piperidine, dimethyl-piperidin-4-yl-amine, pubchem6749, acmc-1akc4, dimethylpiperidin-4-ylamine | |
CN(C)C1CCNCC1 | |
128.22 | |
128.22 |
Chemical Identifiers
50533-97-6 | |
128.22 | |
YFJAIURZMRJPDB-UHFFFAOYSA-N | |
417391 | |
CN(C)C1CCNCC1 |
C7H16N2 | |
MFCD00023144 | |
4-dimethylamino piperidine, 4-dimethylaminopiperidine, 4-n,n-dimethylamino piperidine, 4-piperidinamine, n,n-dimethyl, 4-dimethylamino-piperidine, dimethyl-piperidin-4-yl-amine, pubchem6749, acmc-1akc4, dimethylpiperidin-4-ylamine | |
N,N-dimethylpiperidin-4-amine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 256-617-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only