Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Dimethylamino)piperidine, 97%


Quantity 25g

Manufacturer: alfa aesar™  L2017614

 View more versions of this product

Catalog No. AAL2017614

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 128.22
CAS 50533-97-6
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-(Dimethylamino)piperidine
Assay 97%
Molecular Formula C7H16N2
MDL Number MFCD00023144
Beilstein 1190
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Boiling Point 187°C
UN Number UN2735
Quantity 25g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Specifications

Chemical Name or Material 4-(Dimethylamino)piperidine
CAS 50533-97-6
Molecular Formula C7H16N2
Formula Weight 128.22
MDL Number MFCD00023144
UN Number UN2735
Beilstein 1190

Applications
4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.

Notes
Incompatible with air, water, oxidizing agents and heat.

Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents