missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Dimethylamino)benzonitrile, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 115780250

 View more versions of this product

Catalog No. AC115780250



Specifications

Specifications

4-(Dimethylamino)benzonitrile
Glass bottle
Brown
25g
1197-19-9
100.0
C9H10N2
MFCD00001815
4-cyano-n,n-dimethylaniline
CN(C)C1=CC=C(C=C1)C#N
146.19
146.19
95%
Authentic
318.0°C
70.0°C to 76.0°C
95%
94.0
95%
(CH3)2NC6H4CN
14, 428
JYMNQRQQBJIMCV-UHFFFAOYSA-N
4-(dimethylamino)benzonitrile
70967
Crystalline Powder

Chemical Identifiers

1197-19-9
146.19
JYMNQRQQBJIMCV-UHFFFAOYSA-N
70967
CN(C)C1=CC=C(C=C1)C#N
C9H10N2
MFCD00001815
4-cyano-n,n-dimethylaniline
4-(dimethylamino)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get

GHS Signal Word: Warning

EINECSNumber : 214-819-8

SDS Documents