Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dimethylamino-4'-methylazobenzene, 98%

$39.03 - $142.24

Chemical Identifiers

Chemical Name or Material 4-Dimethylamino-4'-methylazobenzene
CAS 3010-57-9
Molecular Formula C15H17N3
Formula Weight 239.32
MDL Number MFCD00048015
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1857503

  SDS
alfa aesar™
A1857503
1g Each for $39.03

AAA1857506

  SDS
alfa aesar™
A1857506
5g Each for $142.24
Description & Specifications

Specifications

Formula Weight 239.32
Chemical Name or Material 4-Dimethylamino-4'-methylazobenzene
Assay 98%
Molecular Formula C15H17N3
Melting Point 165° to 167°C
MDL Number MFCD00048015
CAS 3010-57-9
Quantity 1g
Odor Odorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.