Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Dimethylamino-3,5-dinitrobenzoic acid, 98%

$60.80 - $220.12

Chemical Identifiers

Chemical Name or Material 4-Dimethylamino-3,5-dinitrobenzoic acid
CAS 82366-55-0
Molecular Formula C9H9N3O6
Formula Weight 255.19
MDL Number MFCD00024508
Beilstein 3402505
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2175406

  SDS
alfa aesar™
B2175406
5g Each for $60.80

AAB2175414

  SDS
alfa aesar™
B2175414
25g Each for $220.12
Description & Specifications

Specifications

Formula Weight 255.19
Chemical Name or Material 4-Dimethylamino-3,5-dinitrobenzoic acid
Assay 98%
Molecular Formula C9H9N3O6
Melting Point 248° to 250°C
MDL Number MFCD00024508
CAS 82366-55-0
Beilstein 3402505
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.