Learn More
4-Dimethylamino-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2359803
Description
4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N'-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N′-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.
Solubility
Slightly soluble 0.32 g/L at 25°C.
Notes
Air sensitive. Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.
Specifications
4-Dimethylamino-2-methoxybenzaldehyde | |
58°C to 60°C | |
MFCD00151814 | |
2719181 | |
4-dimethylamino-2-methoxybenzaldehyde, 2-methoxy-4-dimethylaminobenzaldehyde, 4-dimethylamino-2-methoxy-benzaldehyde, benzaldehyde, 4-dimethylamino-2-methoxy, 2-methoxy-4-dimethylamino benzaldehyde, benzaldehyde,4-dimethylamino-2-methoxy, 2-methoxy-4-n,n-dimethylamino benzaldehyde | |
HGDRXADJVGVGBC-UHFFFAOYSA-N | |
4-(dimethylamino)-2-methoxybenzaldehyde | |
291350 | |
98% |
84562-48-1 | |
C10H13NO2 | |
1 g | |
Air Sensitive | |
Slightly soluble 0.32g/L at 25°C. | |
CN(C)C1=CC(=C(C=C1)C=O)OC | |
179.219 | |
179.22 |
Chemical Identifiers
84562-48-1 | |
179.219 | |
HGDRXADJVGVGBC-UHFFFAOYSA-N | |
4-(dimethylamino)-2-methoxybenzaldehyde |
C10H13NO2 | |
MFCD00151814 | |
291350 | |
CN(C)C1=CC(=C(C=C1)C=O)OC |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only