Please login to your online account to display your discounted pricing

4-(Dimethylamino)-1,1-dimethoxybut-3-en-2-one, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RF03845DA

 View more versions of this product

Catalog No. RF03845DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 173.21
Quantity 1g
Assay 97%
Packaging Amber Glass Bottle
CAS 67751-23-9
Chemical Name or Material 4-(Dimethylamino)-1, 1-dimethoxybut-3-en-2-one
Assay Percent Range 97%
Molecular Formula C8H15NO3
Linear Formula (CH3)2NCH=CHC(O)CH(OCH3)2

Specifications

Chemical Name or Material 4-(Dimethylamino)-1, 1-dimethoxybut-3-en-2-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H15NO3
Linear Formula (CH3)2NCH=CHC(O)CH(OCH3)2
Formula Weight 173.21