Please login to your online account to display your discounted pricing

4-(Dimethoxymethyl)-2-methylpyrimidine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Dimethoxymethyl)-2-methylpyrimidine
CAS 175277-33-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H12N2O2
Formula Weight 168.19
MDL Number MFCD00085137
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB04845DA

  SDS
fisher scientific company
SPB04845DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.19
Quantity 1g
Assay 97%
CAS Min % 97.0
CAS 175277-33-5
Assay Percent Range 97%
Chemical Name or Material 4-(Dimethoxymethyl)-2-methylpyrimidine
MDL Number MFCD00085137
Packaging Amber Glass Bottle
Molecular Formula C8H12N2O2