Please login to your online account to display your discounted pricing

4-(Dimethoxymethyl)-2-methylpyrimidine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB04845DA

 View more versions of this product

Catalog No. SPB04845DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 168.19
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-(Dimethoxymethyl)-2-methylpyrimidine
Assay Percent Range 97%
CAS 175277-33-5
Packaging Amber Glass Bottle
Molecular Formula C8H12N2O2
MDL Number MFCD00085137

Specifications

Chemical Name or Material 4-(Dimethoxymethyl)-2-methylpyrimidine
CAS 175277-33-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H12N2O2
Formula Weight 168.19
MDL Number MFCD00085137