missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Dihexylamino)benzoic acid, 97%, Maybridge

Chemical Identifiers

CAS 134690-32-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C19 H31 N O2
Linear Formula [CH3(CH2)5]2NC6H4CO2H
Formula Weight 305.46
MDL Number MFCD00173873
Chemical Name or Material 4-(Dihexylamino)benzoic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BTB09858DA SDS Fisher Scientific Company
BTB09858DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

134690-32-7
100.0
C19 H31 N O2
305.46
4-(Dihexylamino)benzoic acid
96.5
97%
[CH3(CH2)5]2NC6H4CO2H
MFCD00173873
97%

Specifications

1g
96.5
Amber Glass Bottle
C19 H31 N O2
305.46
4-(Dihexylamino)benzoic acid
134690-32-7
100.0
97%
[CH3(CH2)5]2NC6H4CO2H
MFCD00173873
97%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning