missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Difluoromethoxy)phenyl isocyanate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC319250010

 View more versions of this product

Catalog No. AC319250010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-(Difluoromethoxy)phenyl isocyanate
Clear colorless
Authentic
98.5
Glass bottle
≥98.5%
C8H5F2NO2
185.13
1.3230g/mL
1g
58417-15-5
100.0
Liquid
99%
F2CHOC6H4NCO
MFCD00673056

Chemical Identifiers

58417-15-5
100.0
C8H5F2NO2
185.13
4-(Difluoromethoxy)phenyl isocyanate
98.5
99%
F2CHOC6H4NCO
MFCD00673056
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning