missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Difluoromethoxy)phenyl isocyanate, 99%, ACROS Organics™
Chemical Identifiers
CAS | 58417-15-5 |
---|---|
Molecular Formula | C8H5F2NO2 |
Molecular Weight (g/mol) | 185.13 |
MDL Number | MFCD00673056 |
InChI Key | XLUSNOUGZQJPRN-UHFFFAOYSA-N |
Synonym | 1-difluoromethoxy-4-isocyanato-benzene, 1-difluoromethoxy-4-isocyanatobenzene, 4-difluoromethoxy benzenisocyanate, 4-difluoromethoxy phenyl isocyanate, 4-difluoromethoxy phenyl isosyanate, 4-difluoromethoxy phenylisocyanate, 4-difluoromethoxy-phenyl isocyanate, 4-difluoromethoxyphenyl isocyanate, acmc-1b0ih, benzene,1-difluoromethoxy-4-isocyanato |
PubChem CID | 2733371 |
IUPAC Name | 1-(difluoromethoxy)-4-isocyanatobenzene |
SMILES | C1=CC(=CC=C1N=C=O)OC(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC319255000
|
Thermo Fisher Scientific
AC319255000 |
500mg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC319250010
|
Thermo Fisher Scientific
AC319250010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
58417-15-5 | |
185.13 | |
XLUSNOUGZQJPRN-UHFFFAOYSA-N | |
2733371 | |
C1=CC(=CC=C1N=C=O)OC(F)F |
C8H5F2NO2 | |
MFCD00673056 | |
1-difluoromethoxy-4-isocyanato-benzene, 1-difluoromethoxy-4-isocyanatobenzene, 4-difluoromethoxy benzenisocyanate, 4-difluoromethoxy phenyl isocyanate, 4-difluoromethoxy phenyl isosyanate, 4-difluoromethoxy phenylisocyanate, 4-difluoromethoxy-phenyl isocyanate, 4-difluoromethoxyphenyl isocyanate, acmc-1b0ih, benzene,1-difluoromethoxy-4-isocyanato | |
1-(difluoromethoxy)-4-isocyanatobenzene |
Specifications
1.3230g/mL | |
Authentic | |
1.4940 to 1.4960 | |
209.0°C | |
500mg | |
98.5 | |
98.5% min. (GC) | |
F2CHOC6H4NCO | |
1-difluoromethoxy-4-isocyanato-benzene, 1-difluoromethoxy-4-isocyanatobenzene, 4-difluoromethoxy benzenisocyanate, 4-difluoromethoxy phenyl isocyanate, 4-difluoromethoxy phenyl isosyanate, 4-difluoromethoxy phenylisocyanate, 4-difluoromethoxy-phenyl isocyanate, 4-difluoromethoxyphenyl isocyanate, acmc-1b0ih, benzene,1-difluoromethoxy-4-isocyanato | |
C1=CC(=CC=C1N=C=O)OC(F)F | |
185.13 | |
185.13 | |
99% |
102°C | |
Glass bottle | |
1.323 | |
Colorless | |
58417-15-5 | |
100.0 | |
C8H5F2NO2 | |
MFCD00673056 | |
XLUSNOUGZQJPRN-UHFFFAOYSA-N | |
1-(difluoromethoxy)-4-isocyanatobenzene | |
2733371 | |
Liquid | |
4-(Difluoromethoxy)phenyl isocyanate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic if inhaled.
May cause allergy or asthma symptoms
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger