Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Difluoromethoxy)benzoic acid, 98+%

$295.32 - $295.32

Chemical Identifiers

Chemical Name or Material 4-(Difluoromethoxy)benzoic acid
CAS 4837-20-1
Molecular Formula C8H6F2O3
Formula Weight 188.13
MDL Number MFCD00221454
Beilstein 2615223
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2355906

  SDS
alfa aesar™
B2355906
5g Each for $295.32
Description & Specifications

Specifications

Formula Weight 188.13
Chemical Name or Material 4-(Difluoromethoxy)benzoic acid
Assay ≥98%
Molecular Formula C8H6F2O3
MDL Number MFCD00221454
CAS 4837-20-1
Beilstein 2615223
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 168° to 171°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.