Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Difluoromethoxy)benzaldehyde, 97%

$47.77 - $183.82

Chemical Identifiers

Chemical Name or Material 4-(Difluoromethoxy)benzaldehyde
CAS 73960-07-3
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD00042252
Beilstein 3049286
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2073903

  SDS
alfa aesar™
B2073903
1g Each for $47.77

AAB2073906

  SDS
alfa aesar™
B2073906
5g Each for $183.82
Description & Specifications

Specifications

Formula Weight 172.13
Molecular Formula C8H6F2O2
CAS 73960-07-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 108°C (226°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
MDL Number MFCD00042252
Assay 97%
Chemical Name or Material 4-(Difluoromethoxy)benzaldehyde
Boiling Point 222°C
Quantity 1g
Beilstein 3049286
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.