Please login to your online account to display your discounted pricing

4-(Difluoromethoxy)aniline, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Difluoromethoxy)aniline
CAS 22236-10-8
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H7F2NO
Linear Formula CHF2OC6H4NH2
Formula Weight 159.13
MDL Number MFCD00085005
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC399910010

  SDS
thermo fisher scientific
AC399910010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399910050

  SDS
thermo fisher scientific
AC399910050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.13
Quantity 1g
Density 1.283g/mL
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 4-(Difluoromethoxy)aniline
Linear Formula CHF2OC6H4NH2
CAS 22236-10-8
Assay Percent Range 98%
Molecular Formula C7H7F2NO
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00085005
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.